Learn More
Waters Corp ssDNA LC-MS Standard, 20-mer, Lyophilized, 10 μg
Catalog No. NC2458848 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ssDNA LC-MS Standard is a lyophilized, 10 μg quantity of a 20-mer single-stranded (ss) DNA (sequence: 5′ to 3′ TAA TAC GAC TCA CTA TAG GG). It is a chemically synthesized standard, LC-confirmed for reproducible quantities, designed for performance checks and system suitability in LC-MS applications.
  • Lyophilized, 10 μg quantity of a 20-mer single-stranded (ss) DNA.
  • Sequence: 5′ to 3′ TAA TAC GAC TCA CTA TAG GG.
  • Chemically synthesized and LC-confirmed for reproducible quantities.
  • Designed for performance checks and system suitability in LC-MS analyses.
  • Pre-packaged oligonucleotide reference material for quick proficiency testing.
  • Supports chromatography methods including IPRP, SEC, HILIC, and AEX.
  • Verifies column/system performance and aids in instrument sensitivity testing.
Catalog No. NC2458848 Supplier Waters Corp Supplier No. 186009451
This ssDNA LC-MS Standard is a lyophilized, 10 μg quantity of a 20-mer single-stranded (ss) DNA (sequence: 5′ to 3′ TAA TAC GAC TCA CTA TAG GG). It is a chemically synthesized standard, LC-confirmed for reproducible quantities, designed for performance checks and system suitability in LC-MS applications.
  • Lyophilized, 10 μg quantity of a 20-mer single-stranded (ss) DNA.
  • Sequence: 5′ to 3′ TAA TAC GAC TCA CTA TAG GG.
  • Chemically synthesized and LC-confirmed for reproducible quantities.
  • Designed for performance checks and system suitability in LC-MS analyses.
  • Pre-packaged oligonucleotide reference material for quick proficiency testing.
  • Supports chromatography methods including IPRP, SEC, HILIC, and AEX.
  • Verifies column/system performance and aids in instrument sensitivity testing.