Learn More
Medchemexpress LLC HY-111425 10mg Medchemexpress, SSE15206 CAS:1370046-40-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11142510MG
Encompass_Preferred
Medchemexpress, HY-111425 10mg SSE15206 CAS:1370046-40-4 SSE15206 is a microtubule polymerization inhibitor (GI50 = 197 nM in HCT116 cells) that overcomes multidrug resistance. Causes aberrant mitosis resulting in G2/M arrest due to incomplete spindle formation in cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8866