Learn More
Perkin Elmer US LLC SSI Nut and Ferrule Kit, 0.0625 in
Catalog No. 502113345
Encompass_Preferred
PerkinElmer offers select SSI fittings for 1/16 in. O.D. tubing in 1/4-28 thread size. This kit includes the following pieces: 5 SSI Nuts, 1/16 inch 10 Ferrules, 1/16 inch I.D.
Catalog No. 50-211-3345 Supplier Perkin Elmer US LLC Supplier No. 02540273
PerkinElmer offers select SSI fittings for 1/16 in. O.D. tubing in 1/4-28 thread size. This kit includes the following pieces: 5 SSI Nuts, 1/16 inch 10 Ferrules, 1/16 inch I.D.