Learn More
Cederlane SSI SALMONELLA O FACTOR 150TES
SDP
Catalog No. NC1602623
Encompass
(Product: O:22),(Reacts with factor:G),(No. of tests: 150),(SSI® SALMONELLA O FACTOR ANTISERA FOR SLIDE AGGLUTINATION)
Catalog No. NC1602623 Supplier Cederlane Supplier No. 40282(SS)
(Product: O:22),(Reacts with factor:G),(No. of tests: 150),(SSI® SALMONELLA O FACTOR ANTISERA FOR SLIDE AGGLUTINATION)