Learn More
MSC SSI Schaefer Storage Tote, 55 Lb Capacity, Polypropylene, Gray, Stacking
Catalog No. 5000527100
Encompass
This storage tote is designed for efficient organization and transport of goods. Constructed from durable polypropylene, it offers a 55 lb load capacity and features a stacking design, making it suitable for various industrial and laboratory environments.
  • Load capacity of 55 lb
  • Made from polypropylene
  • Stacking design
  • Non-nesting
  • Gray color
  • Volume capacity of 2.96 cu. ft.
  • Bottom inside width of 13.80 in
  • Bottom inside length of 21.70 in
  • Solid bottom construction
  • FDA approved
Catalog No. 50-005-27100 Supplier MSC Supplier No. 09052366
This storage tote is designed for efficient organization and transport of goods. Constructed from durable polypropylene, it offers a 55 lb load capacity and features a stacking design, making it suitable for various industrial and laboratory environments.
  • Load capacity of 55 lb
  • Made from polypropylene
  • Stacking design
  • Non-nesting
  • Gray color
  • Volume capacity of 2.96 cu. ft.
  • Bottom inside width of 13.80 in
  • Bottom inside length of 21.70 in
  • Solid bottom construction
  • FDA approved