Learn More
Electron Microscopy Sciences PremaDisk S-SiC Grinding Disc P800, 250 mm diameter, 800µm Particles
SDP
Catalog No. 501930271
Encompass_Preferred
Grinding disks made with silicon carbide grains with a 60 percent SiC concentration that is hard resin bonded. The disc can be used in wet or dry applications and have a consistent stock removal rate. They are designed with a flat grain curve that will not flatten after a few applications. They are a good alternative to SiC abrasive paper and SiC grinding foils (films), lasting 50-100 times longer.
Grinding disks come with self-adhesive backing. The discs have a thickness of 0.4 mm.
Diameter: 250 mm
Particles: 800µm
Catalog No. 50-193-0271 Supplier Electron Microscopy Sciences Supplier No. 5029111
Grinding disks made with silicon carbide grains with a 60 percent SiC concentration that is hard resin bonded. The disc can be used in wet or dry applications and have a consistent stock removal rate. They are designed with a flat grain curve that will not flatten after a few applications. They are a good alternative to SiC abrasive paper and SiC grinding foils (films), lasting 50-100 times longer.
Grinding disks come with self-adhesive backing. The discs have a thickness of 0.4 mm.
Diameter: 250 mm
Particles: 800µm