Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ssk36 (TFA) | 99.3% | 1771.04 (free base) | 25 MG
SDP

Catalog No. 5000424892
Encompass_Preferred

ssK36 TFA is a supersubstrate peptide designed for the histone methyltransferase (SET) domain protein 2 (SETD2). It adds methyl groups to the 36th lysine residue of histone H3 (H3K36) to form H3K36me3. This peptide can be methylated by SETD2 over 100 times faster than the natural substrate H3K36, making it valuable for studying the catalytic mechanism of PKMTs, particularly substrate specificity and catalytic efficiency.

  • Designed for the SETD2 protein, a specific PKMT.
  • Adds methyl groups to H3K36 to form H3K36me3.
  • Methylated over 100 times faster than the natural substrate H3K36.
  • Used to study the catalytic mechanism of protein lysine methyltransferases (PKMTs).
  • Suitable for investigating substrate specificity and catalytic efficiency.

Catalog No. 50-004-24892 Supplier Medchemexpress LLC Supplier No. 25MGHYP10463A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ssK36 TFA is a supersubstrate peptide designed for the histone methyltransferase (SET) domain protein 2 (SETD2). It adds methyl groups to the 36th lysine residue of histone H3 (H3K36) to form H3K36me3. This peptide can be methylated by SETD2 over 100 times faster than the natural substrate H3K36, making it valuable for studying the catalytic mechanism of PKMTs, particularly substrate specificity and catalytic efficiency.

  • Designed for the SETD2 protein, a specific PKMT.
  • Adds methyl groups to H3K36 to form H3K36me3.
  • Methylated over 100 times faster than the natural substrate H3K36.
  • Used to study the catalytic mechanism of protein lysine methyltransferases (PKMTs).
  • Suitable for investigating substrate specificity and catalytic efficiency.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.