Learn More
Cayman Chemical SSLV 319 Neurochemicals
Supplier: Cayman Chemical 100090225 MG
A potent and selective CB1 receptor antagonist (Kis = 7.8 and 7,943 nM for CB1 and CB2, respectively); less lipophilic (log P = 5.1) and therefore more water soluble than other known CB1 receptor ligands