Learn More
Cayman Chemical (S)-SLV 319, Purity: ≥98%, CAS: 464213-10-3, 1 mg
SDP
Catalog No. 502888199
Encompass_Preferred
A potent and selective CB1 receptor antagonist (Kis = 7.8 and 7,943 nM for CB1 and CB2, respectively); less lipophilic (log P = 5.1) and therefore more water soluble than other known CB1 receptor ligands
Catalog No. 50-288-8199 Supplier Cayman Chemical Supplier No. 100090221 MG
A potent and selective CB1 receptor antagonist (Kis = 7.8 and 7,943 nM for CB1 and CB2, respectively); less lipophilic (log P = 5.1) and therefore more water soluble than other known CB1 receptor ligands