Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ssm spooky toxin TFA | 99.1% | 6017.84 (free base) | 1 MG
SDP

Catalog No. 5000397076
Encompass_Preferred

Ssm Spooky Toxin TFA is a compound found in *Scolopendra mutilans* that acts as a potent inhibitor of KCNQ (voltage-gated potassium channel family 7) channels. It has IC50s of 2.8 μM, 5.26 μM, and 0.1-0.3 M for Kv7.4, Kv1.3, and Shal channels, respectively. This toxin also inhibits cytokine generation by specifically targeting the KV1.3 channel in T cells and plays an essential role in the centipede's circulatory system.

  • Found in *Scolopendra mutilans*
  • Potently inhibits KCNQ channels
  • Inhibits cytokine generation
  • Plays an essential role in the centipede's circulatory system
  • Molecular weight: 6017.84 (free base)
  • Purity: 99.09%
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (100 mg/mL, requires ultrasonic)
  • Powder storage: 2 years at -80°C, 1 year at -20°C (sealed, away from moisture)
  • In solvent storage: 6 months at -80°C, 1 month at -20°C (sealed, away from moisture)

Catalog No. 50-003-97076 Supplier Medchemexpress LLC Supplier No. HYP5925A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ssm Spooky Toxin TFA is a compound found in *Scolopendra mutilans* that acts as a potent inhibitor of KCNQ (voltage-gated potassium channel family 7) channels. It has IC50s of 2.8 μM, 5.26 μM, and 0.1-0.3 M for Kv7.4, Kv1.3, and Shal channels, respectively. This toxin also inhibits cytokine generation by specifically targeting the KV1.3 channel in T cells and plays an essential role in the centipede's circulatory system.

  • Found in *Scolopendra mutilans*
  • Potently inhibits KCNQ channels
  • Inhibits cytokine generation
  • Plays an essential role in the centipede's circulatory system
  • Molecular weight: 6017.84 (free base)
  • Purity: 99.09%
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (100 mg/mL, requires ultrasonic)
  • Powder storage: 2 years at -80°C, 1 year at -20°C (sealed, away from moisture)
  • In solvent storage: 6 months at -80°C, 1 month at -20°C (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.