Learn More
Bio Rad Laboratories SsoAdvanced Universal SYBR Green Supermix, 500 x 20 uL rxns, 5 mL
Catalog No. NC1031075
Encompass
5 ml (5 x 1 ml vials), 2x qPCR mix, contains dNTPs, Sso7d fusion polymerase, MgCl2, SYBR Green I, ROX Normalization Dyes, for 500 x 20 uL reactions.
Catalog No. NC1031075 Supplier Bio Rad Laboratories Supplier No. 1725271
5 ml (5 x 1 ml vials), 2x qPCR mix, contains dNTPs, Sso7d fusion polymerase, MgCl2, SYBR Green I, ROX Normalization Dyes, for 500 x 20 uL reactions.