Learn More
Bio Rad Laboratories SsoAdvanced Universal SYBR Green Supermix, 2 ml (2 x 1 ml vials), 200 x 20 μl reactions, 2x qPCR mix, Sso7d fusion protein technology
Catalog No. NC1974771
Encompass
SsoAdvanced Universal SYBR Green Supermix, 2 ml (2 x 1 ml vials), 200 x 20 μl reactions, 2x qPCR mix, Sso7d fusion protein technology
Catalog No. NC1974771 Supplier Bio Rad Laboratories Supplier No. 1725270
SsoAdvanced Universal SYBR Green Supermix, 2 ml (2 x 1 ml vials), 200 x 20 μl reactions, 2x qPCR mix, Sso7d fusion protein technology