Learn More
Research Products International Corp SSPE BUFFER, 10 LITER, 20X CONCENTRATED RATE POWDER BLEND, NO LIQUID
SDP
Catalog No. 50213670
Encompass_Preferred
Dry homogeneous blend of molecular biology grade sodium chloride, sodium phosphate and EDTA disodium salt. For use in nucleic acid hybridization at proper working concentration.
Catalog No. 50-213-670 Supplier Research Products International Corp Supplier No. S2404010.0
Dry homogeneous blend of molecular biology grade sodium chloride, sodium phosphate and EDTA disodium salt. For use in nucleic acid hybridization at proper working concentration.