Learn More
Sigma Aldrich Fine Chemicals Biosciences SSPE Buffer 20x Concentrat1L
Catalog No. 502900665
Encompass
SSPE Buffer 20x liquid is a standard reagent in Southern and Northern hybridization procedures. This liquid can be used straight or diluted as needed.
Catalog No. 50-290-0665 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S20151L
SSPE Buffer 20x liquid is a standard reagent in Southern and Northern hybridization procedures. This liquid can be used straight or diluted as needed.