Learn More
Bioworld SSPE Buffer (20X), 4 L
Catalog No. 50196851
Encompass_Preferred
Saline-Sodium Phosphate-EDTA Buffer SSPE 20X is a solution formulated for use in nucleic acid hybridization applications. Technical Information:Each liter prepares 20L of 1X Saline-Sodium Phosphate-EDTA Buffer. Ultra Pure Grade0.22 micron filteredpH @ 25C: 7.25-7.55Storage: RT
Catalog No. 50-196-851 Supplier Bioworld Supplier No. 107500242
Saline-Sodium Phosphate-EDTA Buffer SSPE 20X is a solution formulated for use in nucleic acid hybridization applications. Technical Information:Each liter prepares 20L of 1X Saline-Sodium Phosphate-EDTA Buffer. Ultra Pure Grade0.22 micron filteredpH @ 25C: 7.25-7.55Storage: RT