Learn More
Alkali Scientific SSPE Buffer, 20X Powder Blend, 2103.6 Grams Makes, 10 L
SDP
Catalog No. 5000661153
Encompass_Preferred
SSPE Buffer 20X Powder Blend is designed to prepare a stable 10L solution used in nucleic acid processing applications. This powder blend contains a high concentration of salts to maintain a consistent ionic strength and pH environment. The buffer is primarily used in hybridization assays, washing protocols for membrane filters, and other nucleic acid separation procedures where maintaining a stable ionic concentration and pH is crucial. The 20X concentration allows for easy dilution to the desired working solution. SSPE is ideal for use in molecular biology and genomics research, including Southern and Northern blotting techniques, ensuring proper probe binding and reducing background noise.
Catalog No. 50-006-61153 Supplier Alkali Scientific Supplier No. SSP002CP
SSPE Buffer 20X Powder Blend is designed to prepare a stable 10L solution used in nucleic acid processing applications. This powder blend contains a high concentration of salts to maintain a consistent ionic strength and pH environment. The buffer is primarily used in hybridization assays, washing protocols for membrane filters, and other nucleic acid separation procedures where maintaining a stable ionic concentration and pH is crucial. The 20X concentration allows for easy dilution to the desired working solution. SSPE is ideal for use in molecular biology and genomics research, including Southern and Northern blotting techniques, ensuring proper probe binding and reducing background noise.