Learn More
Bioworld SSPE Buffer, Quick-Mix Pack (20X), 4 PK(s)
Catalog No. 50196853
Encompass_Preferred
Saline-Sodium Phosphate-EDTA Buffer SSPE 20X is a solution formulated for use in nucleic acid hybridization applications. Technical Information:Each Pack prepares 20L of 1X Saline-Sodium Phosphate-EDTA Buffer.Ultra Pure GradepH @ 25C: 7.25-7.55Storage: RTHazard: Irritant
Catalog No. 50-196-853 Supplier Bioworld Supplier No. 107500252
Saline-Sodium Phosphate-EDTA Buffer SSPE 20X is a solution formulated for use in nucleic acid hybridization applications. Technical Information:Each Pack prepares 20L of 1X Saline-Sodium Phosphate-EDTA Buffer.Ultra Pure GradepH @ 25C: 7.25-7.55Storage: RTHazard: Irritant