Learn More
Research Products International Corp RPI SSPE Buffer 20X Solution pH 7.4, 1 Liter
SDP
Catalog No. 501978379
Encompass_Preferred
4X Formulation contains: Sodium Phosphate: 0.2M, Sodium Chloride: 3.0M, EDTA: 20M
Catalog No. 50-197-8379 Supplier Research Products International Corp Supplier No. S241001000.0
4X Formulation contains: Sodium Phosphate: 0.2M, Sodium Chloride: 3.0M, EDTA: 20M