Learn More
Bioworld SSPE Hybridization Solution (6X), 100 mL
Catalog No. 50196854
Encompass_Preferred
SSPE Hybridization Solution SSPE solution is used for the hybridization of probes to nucleic acids Technical Information:SSPE (6X) Blotto (5%)
Catalog No. 50-196-854 Supplier Bioworld Supplier No. 107500271
SSPE Hybridization Solution SSPE solution is used for the hybridization of probes to nucleic acids Technical Information:SSPE (6X) Blotto (5%)