Learn More
Bioworld SSPE Prehybridization Solution (6X), 50 mL
Catalog No. 50196856
Encompass_Preferred
SSPE Prehybridization Solutions SSPE solution is used for the hybridization of probes to nucleic acids Technical Information:SSC (6X) Denhartdt's solution (5X) SDS (0.5%) Denatured, phenol-chloroform extracted from salmon testes DNA (100ug/ml) SSPE (6X)
Catalog No. 50-196-856 Supplier Bioworld Supplier No. 107500281
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SSPE Prehybridization Solutions SSPE solution is used for the hybridization of probes to nucleic acids Technical Information:SSC (6X) Denhartdt's solution (5X) SDS (0.5%) Denatured, phenol-chloroform extracted from salmon testes DNA (100ug/ml) SSPE (6X)