Learn More
Bioworld SSPE Prehybridization Solution (6X), 100 mL
Catalog No. 50196857
Encompass_Preferred
SSPE Prehybridization Solutions SSPE solution is used for the hybridization of probes to nucleic acids Technical Information:SSC (6X) Denhartdt's solution (5X) SDS (0.5%) Denatured, phenol-chloroform extracted from salmon testes DNA (100ug/ml) SSPE (6X)
Catalog No. 50-196-857 Supplier Bioworld Supplier No. 107500282
SSPE Prehybridization Solutions SSPE solution is used for the hybridization of probes to nucleic acids Technical Information:SSC (6X) Denhartdt's solution (5X) SDS (0.5%) Denatured, phenol-chloroform extracted from salmon testes DNA (100ug/ml) SSPE (6X)