Learn More
Inter Dyne Systems Inc Stainless Steel Pegboard, 24" x 36", 40pegs
Catalog No. NC1555652
Encompass_Preferred
V Series pegboards have acatch drain and integral driptrough in 2" or 4" depth. Alldrip troughs some with a36" clear flexible drain tube.• 24"w x 36"h x 3/4"d• 40-6" long 1/2" diameterwhite Polypropylene pegs• Autoclavable• Constructed from 20gauge, type 304 stainlesssteel, polished to a #4 finish• Wall mount bracket shippedwith each pegboard
Catalog No. NC1555652 Supplier Inter Dyne Systems Inc Supplier No. V2436
Add to Cart
Add to Cart
V Series pegboards have acatch drain and integral driptrough in 2" or 4" depth. Alldrip troughs some with a36" clear flexible drain tube.• 24"w x 36"h x 3/4"d• 40-6" long 1/2" diameterwhite Polypropylene pegs• Autoclavable• Constructed from 20gauge, type 304 stainlesssteel, polished to a #4 finish• Wall mount bracket shippedwith each pegboard