Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ssphos

Catalog No. 677280500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-728-0500MG 500 mg
67-728-010G 10 g
67-728-02G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 67-728-0500MG Supplier Sigma Aldrich Supplier No. 677280500MG
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sulfonated ligand for palladium-catalyzed Suzuki-Miyaura coupling under aqueous conditions facilitating isolation of the organic products.Usage subject to US Patents 6307087 and 6395916.

Specifications

CAS 1049726-96-6
Molecular Formula C26H34NaO5PS -+ xH2O
Linear Formula C26H34NaO5PS -+ xH2O
MDL Number MFCD07781994
Quantity 500 mg
Synonym SPhos (water soluble); Sodium 2'-dicyclohexylphosphino-2,6-dimethoxy-1,1'-biphenyl-3-sulfonate hydrate; [1,1'-Biphenyl]-3-sulfonic acid, 2'-(dicyclohexylphosphino)-2,6-dimethoxy- sodium salt
Molecular Weight (g/mol) 512.57 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.