Learn More
TARGETMOL CHEMICALS INC SSR126768A 5MG
SDP
Catalog No. 502580488
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. SSR126768A is a novel and selective orally active and potent oxytocin (OT) receptor antagonist with inhibitory effects on uterine contractions for the prevention of preterm labor. purity: 98%
Catalog No. 50-258-0488 Supplier TARGETMOL CHEMICALS INC Supplier No. T288555MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. SSR126768A is a novel and selective orally active and potent oxytocin (OT) receptor antagonist with inhibitory effects on uterine contractions for the prevention of preterm labor. purity: 98%