Learn More
TARGETMOL CHEMICALS INC SSR128129E 25MG
SDP
Catalog No. 502579581
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SSR128129E (SSR) is an allosteric and orally-active FGFR1 inhibitor (IC50 1.9 uM) but not affecting other related RTKs. purity: 99%
Catalog No. 50-257-9581 Supplier TARGETMOL CHEMICALS INC Supplier No. T608225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SSR128129E (SSR) is an allosteric and orally-active FGFR1 inhibitor (IC50 1.9 uM) but not affecting other related RTKs. purity: 99%