Learn More
Medchemexpress LLC Ssr180711 (hydrochloride) | 446031-79-4 | 99.9% | 361.66 | 100 MG
SDP
Catalog No. 5000384011
Encompass_Preferred
SSR180711 hydrochloride is an orally active, selective, and reversible α7 acetylcholine nicotinic receptor (n-AChRs) partial agonist. It acts on rat α7 n-AChR (Ki=22 nM; IC50=30 nM) and human α7 n-AChR (Ki=14 nM; IC50=18 nM). This compound increases glutamatergic neurotransmission, ACh release, and long-term potentiation (LTP) in the hippocampus. It is intended for research use only.
  • Orally active
  • Selective and reversible α7 n-AChRs partial agonist
  • Increases glutamatergic neurotransmission, ACh release, and long-term potentiation (LTP) in the hippocampus
  • Selective for the α7 receptor subtype compared to α4β2, α3β2, α3β4, and α1β1γδ human n-AChR subtypes (IC50>5 μM)
  • Rapidly penetrates into the brain (ID50=8 mg/kg; p.o.)
Catalog No. 50-003-84011 Supplier Medchemexpress LLC Supplier No. HY19411100MG
SSR180711 hydrochloride is an orally active, selective, and reversible α7 acetylcholine nicotinic receptor (n-AChRs) partial agonist. It acts on rat α7 n-AChR (Ki=22 nM; IC50=30 nM) and human α7 n-AChR (Ki=14 nM; IC50=18 nM). This compound increases glutamatergic neurotransmission, ACh release, and long-term potentiation (LTP) in the hippocampus. It is intended for research use only.
  • Orally active
  • Selective and reversible α7 n-AChRs partial agonist
  • Increases glutamatergic neurotransmission, ACh release, and long-term potentiation (LTP) in the hippocampus
  • Selective for the α7 receptor subtype compared to α4β2, α3β2, α3β4, and α1β1γδ human n-AChR subtypes (IC50>5 μM)
  • Rapidly penetrates into the brain (ID50=8 mg/kg; p.o.)