Learn More
NEXPRING HEALTH SSS-NX (Serum Substitute Supplement – NX) 100ml, Human Serum Albumin (HSA) and alpha (α) and beta (β) globulin culture supplement for gamete and embryo manipulation.
Catalog No. NC2963154
Encompass
SSS-NX (Serum Substitute Supplement-NX) consists of human serum albumin and human serum globulins (α and β) in saline solution and is intended for use in assisted reproductive procedures which include gamete and embryo manipulation and culture. These procedures include the use of SSS-NX as a supplement for culture medium.
This item is not returnable. View return policy
Catalog No. NC2963154 Supplier NEXPRING HEALTH Supplier No. 90194100ML
SSS-NX (Serum Substitute Supplement-NX) consists of human serum albumin and human serum globulins (α and β) in saline solution and is intended for use in assisted reproductive procedures which include gamete and embryo manipulation and culture. These procedures include the use of SSS-NX as a supplement for culture medium.