Learn More
Bellco Glass, Inc. Stainless Steel 1/4 Port Plug For 3L & 7L Bioreactor Systems
SDP
Catalog No. 501944716
Encompass_Preferred
Stainless steel port plug fits onto stainless steel head plate for 3 & 7 liter bioreactor vessel. MADE IN THE USA Material: 316l SST with electropolish finish 1/2-20 tpi threads For 3l & 7l bioreactor systems - English threads For 1/4" port on headplate
Catalog No. 50-194-4716 Supplier Bellco Glass, Inc. Supplier No. 196401014
Stainless steel port plug fits onto stainless steel head plate for 3 & 7 liter bioreactor vessel. MADE IN THE USA Material: 316l SST with electropolish finish 1/2-20 tpi threads For 3l & 7l bioreactor systems - English threads For 1/4" port on headplate