Learn More
Bellco Glass, Inc. 2-Port Sampler Assembly , 45mm
SDP
Catalog No. 501945779
Encompass_Preferred
For use with all vessels having 45mm sidearms. Inner tubes extent 1" into vessel and assembly includes a 45mm blue polypropylene open top cap and white silicone gasket. MADE IN THE USA 316l stainless steel tubes Ra 32 finish or better Polypropylene screw cap
Catalog No. 50-194-5779 Supplier Bellco Glass, Inc. Supplier No. 563721000R
For use with all vessels having 45mm sidearms. Inner tubes extent 1" into vessel and assembly includes a 45mm blue polypropylene open top cap and white silicone gasket. MADE IN THE USA 316l stainless steel tubes Ra 32 finish or better Polypropylene screw cap