Learn More
Bellco Glass, Inc. Stainless Steel 2-Port Assembly , 500mL,70mm For Angled Sidearms
SDP
Catalog No. 501944997
Encompass_Preferred
All port assemblies are manufactured from 316l stainless steel and have an electropolish finish. 2- port assemblies have siphon and vent tubes. 3-port assemblies have siphon, fill tube and vent tubes. All port assemblies include a 45mm blue polypropylene cap and gasket. MADE IN THE USA. For 500mL flasks. Longest tube length 2.5".
Catalog No. 50-194-4997 Supplier Bellco Glass, Inc. Supplier No. 196521005R
All port assemblies are manufactured from 316l stainless steel and have an electropolish finish. 2- port assemblies have siphon and vent tubes. 3-port assemblies have siphon, fill tube and vent tubes. All port assemblies include a 45mm blue polypropylene cap and gasket. MADE IN THE USA. For 500mL flasks. Longest tube length 2.5".