Learn More
Bellco Glass, Inc. Stainless Steel 2-Port Assembly , 5L For Media Bottles
SDP
Catalog No. 501945777
Encompass_Preferred
These stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media or liquids. The 2- port models feature a siphon vent tube and is supplied with a 45mm polypropylene blue open top screw cap and white silicone gasket. MADE IN THE USA
Catalog No. 50-194-5777 Supplier Bellco Glass, Inc. Supplier No. 563720005R
These stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media or liquids. The 2- port models feature a siphon vent tube and is supplied with a 45mm polypropylene blue open top screw cap and white silicone gasket. MADE IN THE USA