Learn More
Bellco Glass, Inc. Stainless Steel 3-Port Assembly ,15L For Angled Sidearms
SDP
Catalog No. 501945007
Encompass_Preferred
Size: 15lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4"OD x .152" IDLongest tube: 11 15/16" at curve. MADE IN THE USA.
Catalog No. 50-194-5007 Supplier Bellco Glass, Inc. Supplier No. 196530015R
Size: 15lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4"OD x .152" IDLongest tube: 11 15/16" at curve. MADE IN THE USA.