Learn More
Bellco Glass, Inc. Stainless Steel 3-Port Assembly , 1L For Media Bottles
SDP
Catalog No. 501945781
Encompass_Preferred
316L stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media. 3-port models feature siphon, fill and vent tubes and is supplied with a 45mm polypropylene blue open top screw cap and white silicone gasket. MADE IN THE USA Dimensions:cap slug: 45mmtube specifications: 1/4" od x 5/32" idsiphon (long tube): 7 19/64" long below cap slugfill tube: 3" long below cap slugvent tube: 1" long below cap slug
Catalog No. 50-194-5781 Supplier Bellco Glass, Inc. Supplier No. 563730001R
316L stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media. 3-port models feature siphon, fill and vent tubes and is supplied with a 45mm polypropylene blue open top screw cap and white silicone gasket. MADE IN THE USA Dimensions:cap slug: 45mmtube specifications: 1/4" od x 5/32" idsiphon (long tube): 7 19/64" long below cap slugfill tube: 3" long below cap slugvent tube: 1" long below cap slug