Learn More
Bellco Glass, Inc. Stainless Steel 3-Port Assembly , 36L
SDP
Catalog No. 501945008
Encompass_Preferred
Size: 36lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4"OD x .152" IDLongest tube: 12 1/8" at bend. MADE IN THE USA.
Catalog No. 50-194-5008 Supplier Bellco Glass, Inc. Supplier No. 196530036R
Size: 36lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4"OD x .152" IDLongest tube: 12 1/8" at bend. MADE IN THE USA.