Learn More
Bellco Glass, Inc. Stainless Steel 3- Port Assembly , 3L For Angled 45mm Sidearms
SDP
Catalog No. 501945003
Encompass_Preferred
Size: 3lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketLongest tube: 5" . MADE IN THE USA.
Catalog No. 50-194-5003 Supplier Bellco Glass, Inc. Supplier No. 196530003R
Size: 3lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketLongest tube: 5" . MADE IN THE USA.