Learn More
Bellco Glass, Inc. Stainless Steel 3-Port Assembly ,500mL 70mm
SDP
Catalog No. 501945012
Encompass_Preferred
Size: 500mlSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4" x 5/32"Longest tube: 2 5/8" at bendNeck: 70mm. MADE IN THE USA.
Catalog No. 50-194-5012 Supplier Bellco Glass, Inc. Supplier No. 196531005R
Size: 500mlSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4" x 5/32"Longest tube: 2 5/8" at bendNeck: 70mm. MADE IN THE USA.