Learn More
Bellco Glass, Inc. Stainless Steel 3-Port Assembly , 5L For Media Bottles
SDP
Catalog No. 501945783
Encompass_Preferred
316L stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media. 3-port models feature siphon, fill and vent tubes and is supplied with a 45mm polypropylene blue open top screw cap and white silicone gasket. MADE IN THE USADimensions:cap slug: 45mmtube specifications: 1/4″ od x 5/32″ idsiphon (long tube): 11 7/16″ long below cap slugfill tube: 3 ” long below cap slugvent tube: 1″ long below cap slug
Catalog No. 50-194-5783 Supplier Bellco Glass, Inc. Supplier No. 563730005R
316L stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media. 3-port models feature siphon, fill and vent tubes and is supplied with a 45mm polypropylene blue open top screw cap and white silicone gasket. MADE IN THE USADimensions:cap slug: 45mmtube specifications: 1/4″ od x 5/32″ idsiphon (long tube): 11 7/16″ long below cap slugfill tube: 3 ” long below cap slugvent tube: 1″ long below cap slug