Learn More
Bellco Glass, Inc. Stainless Steel 3-Port Assembly , 6L For Angled Sidearms
SDP
Catalog No. 501945005
Encompass_Preferred
Size: 6lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4" OD x .152" IDLongest tube: 10 1/2" at curve. MADE IN THE USA.
Catalog No. 50-194-5005 Supplier Bellco Glass, Inc. Supplier No. 196530006R
Size: 6lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4" OD x .152" IDLongest tube: 10 1/2" at curve. MADE IN THE USA.