Learn More
Bellco Glass, Inc. Stainless Steel 3-Port Assembly , 8L For Angled Sidearms
SDP
Catalog No. 501945006
Encompass_Preferred
Size: 8lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4" OD x .152" IDLongest tube: 11". MADE IN THE USA.
Catalog No. 50-194-5006 Supplier Bellco Glass, Inc. Supplier No. 196530008R
Size: 8lSeam welded w/ electropolish finish3-port w/ siphon, fill & vent tubesIncludes: 45mm pp cap & silicone gasketDimensions: 1/4" OD x .152" IDLongest tube: 11". MADE IN THE USA.