Learn More
Bellco Glass, Inc. Stainless Steel 5/16OD Cooling Coil, 10L For 10L Bioreactor Systems"
SDP
Catalog No. 501944731
Encompass_Preferred
Heating and cooling coils are used for cell culture temperature manipulation within the bioreactor vessel. Must be hooked up to a water circulating mechanism for heating or cooling. MADE IN THE USA SST cooling coils For 10l bioreactor systems - English threads Tubing Specifications" 5/16" OD Material: 316L SST with Electropolish Finish Overall Length: 17 5/8" OD of Coil: 4 5/8" Number of turns: 5
Catalog No. 50-194-4731 Supplier Bellco Glass, Inc. Supplier No. 196402175
Heating and cooling coils are used for cell culture temperature manipulation within the bioreactor vessel. Must be hooked up to a water circulating mechanism for heating or cooling. MADE IN THE USA SST cooling coils For 10l bioreactor systems - English threads Tubing Specifications" 5/16" OD Material: 316L SST with Electropolish Finish Overall Length: 17 5/8" OD of Coil: 4 5/8" Number of turns: 5