Learn More
Bellco Glass, Inc. Stainless Steel Angled 2-Port Assembly ,45mm With Hose Barbs - RA 32
SDP
Supplier:  Bellco Glass, Inc. 563712000R
Encompass_Preferred
Stainless Steel Angled 2-Port assembly 45mm with Hose Barbs for easy attachment of silicone tubing. 2-Port Assembly attaches to vessel side arms with 45mm screw cap. MADE IN THE USA 316l stainless steel finish of ra 32 or better - inside & out TUBE SPECIFICATIONS: 1/4" OD X .156" ID
Catalog No. 50-194-5774