Learn More
Chemglass Life Sciences 2-Port Angled Vent Assembly, 90° 6.35mm/0.25" OD Hose Barbs, GL-45, Universal 316L Electro Polished Stainless Steel, Complete with Gasket and Cap
SDP
Catalog No. 501214879
Encompass
Universal vent port assembly is constructed of 316L electro polished stainless steel.These port assemblies fit all types of spinner flasks, media bottles and vessels.Configurations consist of a single vent tube, vent and short fill tubes, or angled vent and fill tubes.Supplied complete with a GL-32 or GL-45 open top cap and silicone gasket.
Catalog No. 50-121-4879 Supplier Chemglass Life Sciences Supplier No. CLS142921
Universal vent port assembly is constructed of 316L electro polished stainless steel.These port assemblies fit all types of spinner flasks, media bottles and vessels.Configurations consist of a single vent tube, vent and short fill tubes, or angled vent and fill tubes.Supplied complete with a GL-32 or GL-45 open top cap and silicone gasket.