Learn More
Aviva Systems Biology Corp Sst Antibody - N-terminal region (ARP41905_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41905_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sst. It was validated on Western Blot. . Alternate names: SOM, SRIF, SS, SmstApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-284-965