Learn More
Bellco Glass, Inc. Stainless Steel Baffle Assembly , 10L For 10L Bioreactor Systems
SDP
Catalog No. 501944902
Encompass_Preferred
Stainless Steel Baffle Assembly , 10L For 10L Bioreactor Systems. Manufactured from 316l stainless steel with ra32 finish or better inside and out. The assemblies attach to the headplate. MADE IN THE USA.
Catalog No. 50-194-4902 Supplier Bellco Glass, Inc. Supplier No. 196488610
Stainless Steel Baffle Assembly , 10L For 10L Bioreactor Systems. Manufactured from 316l stainless steel with ra32 finish or better inside and out. The assemblies attach to the headplate. MADE IN THE USA.