Learn More
Bellco Glass, Inc. Stainless Steel Baffle Assembly , 3L For Bioreactor Systems
SDP
Catalog No. 501944903
Encompass_Preferred
Manufactured from 316l stainless steel with ra32 finish or better inside and out. The assemblies attach to the headplate. MADE IN THE USA. SST Baffle AssemblyFor 3L English HeadplatesSize: 3LMaterial: 316L SST with electropolish finishBaffle Shaft: .250" OD x 6 1/4" long (1/4-28 TPI thread)Baffle Plate: .625" OD x 7" long x .090" thickNoInsertion Length: 9 1/8"Supply with A542-407 - O-ring & 1/4-28 SST Hex Nut
Catalog No. 50-194-4903 Supplier Bellco Glass, Inc. Supplier No. 196488630
Manufactured from 316l stainless steel with ra32 finish or better inside and out. The assemblies attach to the headplate. MADE IN THE USA. SST Baffle AssemblyFor 3L English HeadplatesSize: 3LMaterial: 316L SST with electropolish finishBaffle Shaft: .250" OD x 6 1/4" long (1/4-28 TPI thread)Baffle Plate: .625" OD x 7" long x .090" thickNoInsertion Length: 9 1/8"Supply with A542-407 - O-ring & 1/4-28 SST Hex Nut