Learn More
Bellco Glass, Inc. Stainless Steel Bent Harvesting Tube,15L 1/2 OD x 20"L - Flat Bottom Vessel"
SDP
Catalog No. 501944943
Encompass_Preferred
316l stainless steel bent harvesting tube for 15l flat bottom omni vessel. MADE IN THE USA. Use with large flange head-plate. Size: 1/2" x 20" Id: .430"
Catalog No. 50-194-4943 Supplier Bellco Glass, Inc. Supplier No. 196491115
316l stainless steel bent harvesting tube for 15l flat bottom omni vessel. MADE IN THE USA. Use with large flange head-plate. Size: 1/2" x 20" Id: .430"