Learn More
Bellco Glass, Inc. Stainless Steel Bent Harvesting Tube, 15L 1/4OD x 20"L - Ra32・Finish
SDP
Catalog No. 501944931
Encompass_Preferred
316l stainless steel bent harvesting tube for 15l flat bottom omni vessel. Use with large flange head-plate. MADE IN THE USA. Size: 1/4" x 20" Id: 5/32"
Catalog No. 50-194-4931 Supplier Bellco Glass, Inc. Supplier No. 196490080
316l stainless steel bent harvesting tube for 15l flat bottom omni vessel. Use with large flange head-plate. MADE IN THE USA. Size: 1/4" x 20" Id: 5/32"