Learn More
Bellco Glass, Inc. Stainless Steel Bent Harvesting Tube, 36L 1/2OD x 22" L - Flat Bottom Vessel"
SDP
Catalog No. 501944944
Encompass_Preferred
316l stainless steel bent harvesting tube for 36l flat bottom omni vessel. MADE IN THE USA. Use with large flange head-plate. Size: 1/2" x 22" Id: .430"
Catalog No. 50-194-4944 Supplier Bellco Glass, Inc. Supplier No. 196491136
316l stainless steel bent harvesting tube for 36l flat bottom omni vessel. MADE IN THE USA. Use with large flange head-plate. Size: 1/2" x 22" Id: .430"