Learn More
Bellco Glass, Inc. Stainless Steel Bent Harvesting Tube, 5L 1/4OD x 16"L - Ra32・Finish
SDP
Catalog No. 501944929
Encompass_Preferred
316l stainless steel bent harvesting tube for 5l flat bottom omni vessel. Use with large flange head-plate. MADE IN THE USA. Size: 1/4" x 16" Id: 5/32"
Catalog No. 50-194-4929 Supplier Bellco Glass, Inc. Supplier No. 196490070
316l stainless steel bent harvesting tube for 5l flat bottom omni vessel. Use with large flange head-plate. MADE IN THE USA. Size: 1/4" x 16" Id: 5/32"