Learn More
Bellco Glass, Inc. Stainless Steel Bent Harvesting Tube,8L 1/2 OD x 18" - Flat Bottom Vessel"
SDP
Catalog No. 501944942
Encompass_Preferred
SST Bent Harvesting Tube. For 8L Large Flange Omni Vessels. MADE IN THE USA. Material: 316l SST with electropolish finish - outside onlyDimensions: 1/2" od x .430"id x 18" L
Catalog No. 50-194-4942 Supplier Bellco Glass, Inc. Supplier No. 196491108
SST Bent Harvesting Tube. For 8L Large Flange Omni Vessels. MADE IN THE USA. Material: 316l SST with electropolish finish - outside onlyDimensions: 1/2" od x .430"id x 18" L